Learn More
Hach Company EDTA, Tetrasodium Salt, 500 g
Supplier:  Hach Company 700534
Encompass
Ethylenediaminetetraacetic Acid, Tetrasodium Salt Dihydrate, Analytical Grade, CAS No. 64-02-8 C2H4N2(CH2COO)4Na4·...2H2O, FW = 416.23. Assay: 98.0-102.0% Insoluble Matter: ≤0.02%
Catalog No. 50-199-4822