Learn More
Braintree Scientific Minivette POCT, 20ul, EDTA, red, 200/pk
SDP
Supplier:  Braintree Scientific MP502113PK
Encompass
Minivette POCT - Easy sample recovery due to optimal capillary features. Precise dispensing of small whole blood volumes. Prevents spillage during transfer to test card. Available in 20ul and 50ul volumes. Preparations: Neutral, heparin and EDTA **Sold for research purposes only and not for human use**
Catalog No. NC1277110
 Restricted Item