Learn More
Hach Company EDTA Solution, 1M, 50 mL SCDB
Supplier:  Hach Company 2241926
Encompass
For determination of calcium and magnesium hardness in water by the Calmagite Colorimetric method. Hach Method 8030. 50 mL Self-Contained Dropping Bottle.
Catalog No. 50-199-5606