Learn More
Aqua Solutions EDTA TITRANT 0.001 MOLAR(4LC)
SDP
Supplier:  Aqua Solutions 35534LC
Encompass_Preferred
EDTA Titrant, 0.001 Molar APHA for Hardness; <0.1%; Clear odorless liquid; 4L; <5 deg.C; > 95 deg.C; CAS: 6381-92-6; Store in a cool dry place
Catalog No. NC9521116
Only null left
Add to Cart
Description
Description
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov