Learn More
EDTA Titrant, 0.0250 Molar, Reagents
EDTA Titrant, 0.0250 Molar, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.
$64.00
Specifications
| Chemical Name or Material | EDTA Titrant, 0.0250 Molar |
|---|---|
| Color | Colorless |
| CAS | 7732-18-5 , 6381-92-6 |
| Boiling Point | 100°C |
| Molecular Formula | C10H14N2O8Na2/H2O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
01-335-377
|
Reagents Holdings Llc
CE1055001A |
1 L |
Each of 1 for $64.00
|
|
|||||
|
You must Sign In and have a business account to purchase this item. Restricted Item |
|||||||||
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.