Learn More
Apexbio Technology LLC EdU Flow Cytometry Assay Kit (Cy5), 100 tests
SDP
Supplier:  Apexbio Technology LLC K1078100
Encompass_Preferred
EdU Flow Cytometry Assay Kit (Cy5) is a sensitive and reliable way to detect and quantify cell proliferation in mammalian cells. It is optimized for flow cytometry. It is a better alternative to traditional BrdU (bromodeoxyuridine). Other sizes are also available. Please inqury us for quote.
Catalog No. 50-199-8208