Learn More
Medchemexpress LLC HY-101117 10mg Medchemexpress, EED226 CAS:2083627-02-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY10111710MG
Encompass_Preferred
Medchemexpress, HY-101117 10mg EED226 CAS:2083627-02-3 EED226 is a polycomb repressive complex 2 (PRC2) inhibitor, which binds to the K27me3-pocket on embryonic ectoderm development (EED) and shows strong antitumor activity in xenograft mice model. EED226 is a potent, selective, and orally bioavailable EED inhibitor[2]. EED226 inhibits PRC2 with an IC50 of 23.4 nM when the H3K27me0 peptide is used as a substrate in the in vitro enzymatic assays[3]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8647
 Restricted Item