Learn More
Selleck Chemical LLC EED226 5mg 2083627-02-3
SDP
Supplier:  Selleck Chemical LLC S84965MG
Encompass_Preferred
EED226 is a potent, selective, and orally bioavailable a novel allosteric Polycomb repressive complex 2 (PRC2) inhibitor with an IC50 of 23.4 nM when the H3K27me0 peptide was used as substrate and an IC50 of 53.5 nM when the mononucleosome was used as the substrate. It directly binds to the H3K27me3 binding pocket of EED. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-6091