Learn More
Medchemexpress LLC EED226-COOH | 2467965-71-3 | MFCD32701933 | 99.9% | 335.32 g·mol⁻1 | C17H13N5O3 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1309795MG
Encompass_Preferred
EED226-COOH is an EED226-derived small-molecule ligand that binds the EED subunit of polycomb repressive complex 2 (PRC2). Supplied as a research reagent, it is used as an EED-targeting ligand in PROTAC design to enable PRC2 degradation.
  • High purity (99.88%).
  • Molecular formula C17H13N5O3; molar mass 335.32 g·mol⁻1.
  • Used as an EED-targeting ligand in PROTAC construction and degradation studies.
  • Available in small research quantities (e.g., 5 mg).
  • Dissolves in DMSO for assay preparation.
Catalog No. 50-002-62795