Learn More
Amsbio LLC EF1a-Luciferase (firefly)/ (Hygro)
Supplier:  Amsbio LLC LVP1239PBS
Encompass
Lucifierase (firefly) expression lentivirus under the enhanced EF1a promoter, with a Hygromycin antibiotic selection under RSV promoter.
Catalog No. NC2744148