Learn More
Medchemexpress LLC Efaroxan hydrochloride | 89197-00-2 | 99.8% | 5 MG
SDP
Supplier:  Medchemexpress LLC HYB1416A5MG
Encompass_Preferred
Efaroxan hydrochloride is a potent, selective and orally active α2-adrenoceptor antagonist, with antidiabetic activity. It is a selective I1-Imidazoline receptor antagonist and can be used for the research of cardiovascular disease.
  • Potent, selective, and orally active α2-adrenoceptor antagonist.
  • Exhibits antidiabetic activity.
  • Selective I1-Imidazoline receptor antagonist.
  • Potentially useful for research in cardiovascular disease.
  • Increases plasma insulin levels in rats without significantly affecting plasma glucose levels (In Vivo finding).
Catalog No. 50-004-12527