Learn More
Apexbio Technology LLC Efavirenz 154598-52-4 50mg
SDP
Supplier:  Apexbio Technology LLC B111950
Encompass_Preferred
Efavirenz is a non-nucleoside reverse transcriptase inhibitor targeting human immunodeficiency virus type-1 (HIV-1) reverse transcriptase (RT) It functions by binding specifically to an allosteric site of HIV-1 RT thereby inhibiting viral RNA-dependent DNA polymerase activity essential for HIV replication Efavirenz exhibits inhibitory activity against wild-type HIV-1 RT with reported IC95 values ranging from approximately 1 5 to 3 nM and retains inhibitory potency against several known NNRTI-resistant RT variants Due to its molecular mechanism and specificity Efavirenz is frequently utilized in antiviral studies particularly HIV research investigating RT inhibition and HIV resistance mechanisms
Catalog No. 50-000-62322