Learn More
Medchemexpress LLC Efbemalenograstim al 10mg | 10mg
SDP
Supplier:  Medchemexpress LLC HYP9987910MG
Encompass_Preferred
Efbemalenograstim alfa (F 627) is a recombinant fusion protein Efbemalenograstim alfa is a long acting dimeric granulocyte colony-stimulating factor (G-CSF) that contains two human G-CSF fused to a human immunoglobulin G2 (hIgG2)-Fc fragment with a peptide linker Efbemalenograstim alfa induces the production of white blood cells[1]
Catalog No. 50-003-22170