Learn More
Cayman Chemical DL-α-Difluoromethylornithine (hydrochloride hydrate), 96020-91-6, 25 mg
SDP
Supplier:  Cayman Chemical 1688925
Encompass_Preferred
An irreversible inhibitor of ornithine decarboxylase that suppresses polyamine biosynthesis; displays antiangiogenic, cytostatic, and antiparasitic effects
Catalog No. NC1415417