Learn More
Selleck Chemical LLC Efonidipine HCl ethanolate
SDP
Supplier:  Selleck Chemical LLC S589825MG
Encompass_Preferred
Efonidipine (NZ-105) hydrochloride monoethanolate is an L- and T-type calcium channel blocker leading to vasodilation and decreased automaticity of the heart Efonidipine hydrochloride monoethanolate also suppresses aldosterone secretion from the adrenal
Catalog No. 50-000-03545