Learn More
Cayman Chemical EG 00229trIfluoROCTAte sa 10mg
SDP
Supplier:  Cayman Chemical 3393710 MG
Encompass_Preferred
An NRP-1 antagonist; inhibits the binding of VEGF-A to porcine aortic endothelial cells expressing NRP-1 but not VEGFR2 (IC50 = 8 µM); reduces VEGF-A-induced migration of HUVECs and enhances the cytotoxicity of paclitaxel or 5-fluorouracil in A549 lung cancer cells; reduces SARS-CoV-2 infectivity in Caco-2 cells at 100 µM
Catalog No. 50-294-7516