Learn More
Medchemexpress LLC Eg00229 trifluoroacetate | 1210945-69-9 | 98.0% | 611.60 | C19H20F3N7O7S3 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY10799100MG
Encompass_Preferred
EG00229 trifluoroacetate is a small-molecule neuropilin-1 (NRP1) receptor antagonist supplied for research use. It selectively inhibits VEGF-A binding to the NRP1 b1 domain and is intended for in vitro and preclinical studies of NRP1-mediated signaling.
  • Neuropilin-1 receptor antagonist that blocks VEGF-A binding.
  • Reported IC50 ≈3 μM against purified NRP1 b1 domain and ≈8 μM in cell binding assays.
  • Trifluoroacetate salt form for consistent formulation.
  • High purity (≈98.0%).
  • Molecular weight 611.6, suitable for biochemical and binding assays.
  • Intended for research use in mechanistic and signaling studies.
Catalog No. 50-002-42153