Learn More
Medchemexpress LLC Eg01377 dihydrochloride | 2749438-61-5 | MFCD32701908 | 99.0% | 659.60 | C26H32Cl2N6O6S2 | 5 MG

Supplier: Medchemexpress LLC HY112151A5MG
EG01377 dihydrochloride is a selective, bioavailable small-molecule inhibitor of neuropilin-1 (NRP1) for research applications. It exhibits a Kd of 1.32 μM and IC50 values of 609 nM for NRP1-a1 and NRP1-b1, and is provided as a solid suitable for in vitro and preclinical studies.
- Selective neuropilin-1 inhibition with measured potency.
- Demonstrated activity: Kd 1.32 μM; IC50 609 nM.
- Supplied as a white to off-white solid for laboratory use.
- High reported purity (98.98%).
- Molecular weight 659.60; formula C26H32Cl2N6O6S2.
- Recommended storage under inert atmosphere, away from moisture; in solvent: -80°C (6 months), -20°C (1 month).
- Available in small milligram quantities for research workflows.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.