Learn More
Medchemexpress LLC HY-100716 5mg , Eganelisib CAS:1693758-51-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1007165MG
Encompass_Preferred
HY-100716 5mg Eganelisib CAS: 1693758-51-8 Eganelisib (IPI549) is a potent and selective PI3Kγ inhibitor with an IC50 of 16 nM. Eganelisib shows >100-fold selectivity over other lipid and protein kinases. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9418