Learn More
Medchemexpress LLC HY-N6263 5mg Medchemexpress, EGCG Octaacetate CAS:148707-39-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN62635MG
Encompass_Preferred
Medchemexpress, HY-N6263 5mg EGCG Octaacetate CAS:148707-39-5 EGCG Octaacetate is a prodrug of Green tea epigallocatechin-3-gallate (EGCG), utilized to enhance the stability and bioavailability of EGCG in vivo. EGCG Octaacetate has high efficacy, bioavailability, anti-oxidation and anti-angiogenesis capacities. EGCG octaacetate is the potential antibacterial compound for gram-positive bacteria (GPB) and gram-negative bacteria (GNB) . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2951