Learn More
Medchemexpress LLC EGF Canine P.pasto 5ug
SDP
Supplier:  Medchemexpress LLC HYP729805UG
Encompass_Preferred
The EGF protein plays a key role in promoting the growth of a variety of epidermal and epithelial tissues in vivo and in vitro and in stimulating the proliferation of certain fibroblasts in cell culture In addition it also has the effect of magnesium-stimulating hormone causing magnesium reabsorption in the renal distal convoluted tubule through the participation of EGFR and the activation of magnesium channel TRPM6 EGF Protein Canine (P pastoris) is the recombinant canine-derived EGF protein expressed by P pastoris with tag free
Catalog No. 50-001-82760