Learn More
Carbosynth LLC. EGF (Human) (0.1 mg vial), CAS# 62253-63-8, MF= C270H395N73O83S7
SDP
Supplier:  Carbosynth LLC. PGR4498S0.1MG
Encompass_Preferred
This item has a minimum order quantity of 2 per supplier requirements
-Urogastrone, Epidermal Growth Factor (Human); Asn-Ser-Asp-Ser-Glu-Cys-Pro-Leu-Ser-His-Asp-Gly-Tyr-Cys-Leu-His-Asp-Gly-Val-Cys-Met-Tyr-Ile-Glu-Ala-Leu-Asp-Lys-Tyr-Ala-Cys-Asn-Cys-Val-Val-Gly-Tyr-Ile-Gly-Glu-Arg-Cys-Gln-Tyr-Arg-Asp-Leu-Lys-Trp-Trp-Glu-Leu-Arg, Application: Growth Factor that Stimulates Cell Growth, Proliferation, and Differentiation Synthetic Product , Notes: Epidermal Growth Factor (EGF) EGF is a 6-kDa protein consisting of 53 amino acids residues with three intermolecular disulfide bonds. It was originally isolated from the sub-maxillary glands of mice by Cohen. EGF stimulates the cell proliferation and differentiation by binding the receptor EGFR., MW: 6215.9, Disulfide bonds: Cys6-Cys20, Cys14-Cys31, and Cys33-Cys42
Catalog No. 50-195-2418