Learn More
Santa Cruz Biotechnology Anti-EGFR Antibody (A-10), 200 ug/mL
Supplier:  Santa Cruz Biotechnology SC373746
Encompass
Anti-EGFR Antibody (A-10) is recommended for detection of EGFR of mouse, rat and human origin by WB, IP, IF, IHC(P) and ELISA.
Catalog No. NC1848928