Learn More
Sino Biological Recombinant Human EGFR Protein (G719C, GST Tag), Active
Supplier:  Sino Biological E1012FG500
Encompass_Preferred
Recombinant human EGFR (G719C) (695-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
Catalog No. 50-257-5186