Learn More
Medchemexpress LLC EGFR/ErbB-2-IN-2 | 179248-61-4 | 99.5% | 387.43 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY11242050MG
Encompass_Preferred
EGFR/ErbB-2-IN-2 is an EGFR and ErbB inhibitor with IC50s of 0.017 μM, 0.08 μM, and 1.91 μM for EGFR, ErbB-2, and ErbB-4, respectively. It also inhibits c-Src, VEGFR2, and c-Fms with IC50s of 0.30, 2.2, and 14.0 μM, respectively.
  • EGFR and ErbB inhibitor
  • Inhibits EGFR, ErbB-2, ErbB-4, c-Src, VEGFR2, and c-Fms
  • Purity of 99.45%
Catalog No. 50-004-35699