Learn More
Medchemexpress LLC EGFR Human sf9 Hi 5ug
SDP
Supplier:  Medchemexpress LLC HYP729875UG
Encompass_Preferred
EGFR is a transmembrane glycoprotein EGFR binds to ligands triggering receptor dimerization inducing intracellular TK domain autophosphorylation activating downstream Ras/MAPK PI3K/Akt and other signaling pathways and regulating cell proliferation survival migration and angiogenesis EGFR Protein Human (sf9 His-GST) is a recombinant EGFR protein expressed by Sf9 insect cells with N-His N-GST tags
Catalog No. 50-001-80769