Learn More
TARGETMOL CHEMICALS INC EGFR-IN-1 HYDROCHLORIDE 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T11157L5MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. EGFR-IN-1 hydrochloride is an irreversible and specific inhibitor of L858R/T790M mutant EGFR with 100-fold selectivity over wild-type EGFR. EGFR-IN-1 hydrochloride exhibits potent antitumor and antiproliferative activity in H1975 cells and the mutant HCC827 cells with IC50s of 4 and 28 nM respectively. purity: 99%
Catalog No. 50-258-1560