Learn More
Medchemexpress LLC Egfr-in-5 | 2225887-26-1 | 99.5% | 571.69 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY11141510MG
Encompass_Preferred
EGFR-IN-5 is an EGFR inhibitor with IC50s of 10.4, 1.1, 34, and 7.2 nM for EGFR, EGFRL858R, EGFRL858R/T790M, and EGFRL858R/T790M/C797S, respectively. It is for research use only.
  • Appearance: Solid
  • Color: Light yellow to yellow
Catalog No. 50-003-44275