Learn More
Medchemexpress LLC EGFR-IN-69 | 2433837-65-9 | C31H37Cl2N7O3S | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1506105MG
Encompass_Preferred
EGFR-IN-69 (compound 17g) is a potent EGFR inhibitor used in non-small-cell-lung-cancer (NSCLC) research. It demonstrates strong inhibitory activity against multiple EGFR mutations, including EGFRL858R/T790M/C797S (IC50 4.3 nM), EGFRL858R/T790M (IC50 6.6 nM), and EGFR19del/T790M/C797S (IC50 25.6 nM). The compound also shows antiproliferative and cytotoxic effects on BaF3 cell lines.
  • Potent EGFR inhibitor
  • Inhibits EGFR L858R/T790M/C797S, L858R/T790M, and del19/T790M/C797S mutations
  • Used in NSCLC research
  • Antiproliferative and cytotoxic activity against BaF3 cell lines
  • Light yellow to yellow solid
  • 99.58% purity
  • For research use only
Catalog No. 50-003-75102