Learn More
Medchemexpress LLC EGFR-IN-98 | 99.1% | 383.87 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY16112610MM/1ML
Encompass_Preferred
EGFR-IN-98 (Compound 4c) is an EGFR inhibitor that can be used in the study of tumors. It demonstrates potent inhibitory activity against various EGFR mutations. This product is provided as a solid with high purity, suitable for research applications.
  • Targets EGFR, specifically mutations L858R/T790M/C797S and Del19/T790M/C797S
  • High purity of 99.12%
  • Molecular weight of 383.87
  • Available as a solid, or in solution with DMSO
  • Suitable for tumor studies
Catalog No. 50-003-94167