Learn More
Cayman Chemical EGFR InhIbItr 5mg
SDP
Supplier:  Cayman Chemical 153635 MG
Encompass_Preferred
A cell permeable, selective inhibitor of the EGFR kinase (IC50s = 21, 63, and 4 nM for EGFR and the L858R and L861Q EGFR mutants, respectively)
Catalog No. 50-297-1734