Learn More
Medchemexpress LLC EGFR Rat HEK293 H 2ug
SDP
Supplier:  Medchemexpress LLC HYP729902UG
Encompass_Preferred
EGFR Protein is a receptor tyrosine kinase that binds to ligands of the EGF family EGFR Protein is involved in the regulation of NF-kappa-B RAS-RAF-MEK-ERK PI3 kinase-AKT PLCgamma-PKC and STATs signaling pathways EGFR Protein has calmodulin binding activity epidermal growth factor binding activity and epidermal growth factor activating receptor activity EGFR Protein Rat (HEK293 His) is the recombinant rat-derived EGFR protein expressed by HEK293 with C-His labeled tag
Catalog No. 50-001-81021