Learn More
Medchemexpress LLC EGFR T790M/L858R-IN-2 | 2955607-40-4 | 99.7% | 497.57 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY14982450MG
Encompass_Preferred
EGFR T790M/L858R-IN-2 is a potent and selective inhibitor that targets EGFRT790M/L858R with high efficacy. It plays a role in decreasing the expression of p-EGFR, P-AKT, and P-ERK1/2, inducing apoptosis, and promoting cell cycle arrest in the G1 phase.
  • Potent and selective EGFRT790M/L858R inhibitor
  • Decreases expression of p-EGFR, P-AKT, and P-ERK1/2
  • Induces apoptosis
  • Induces cell cycle arrest in the G1 phase
  • Shows anti-cancer activity
Catalog No. 50-003-61844