Learn More
Medchemexpress LLC EGFR-TK-IN-4 10mM | 3053485-72-3 | >99.9% | 510.54 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY16192710MM/1ML
Encompass_Preferred
EGFR-TK-IN-4 (compound 10k) is a potent and selective EGFR-TK inhibitor. It can induce apoptosis and has antitumor activity.
  • Potent and selective EGFR-TK inhibitor.
  • Induces apoptosis.
  • Exhibits antitumor activity.
  • Effective against human A549 cells with an IC50 of 0.82 μM.
  • Shows cytotoxicity against human BEAS-2B cells with an IC50 of 38.41 μM.
  • Demonstrates anticancer activity against human HCC827 cells with an IC50 of 0.01 μM.
  • Active against human NCI-H1975 cells with an IC50 of 0.08 μM.
Catalog No. 50-003-94930