Learn More
Cayman Chemical EGFRCTerm RabbIt Monclon 100uL
SDP
Supplier:  Cayman Chemical 32235100 UL
Encompass_Preferred
A PKC inhibitor (IC50s 1.9 and 0.4 nM for PKCa and PKCθ respectively) selective for PKCa and PKCθ over GSK3B (IC50 3100 nM) inhibits proliferation of TMD8 B cell lymphoma and 92.1 uveal melanoma cells (IC50s 900 and 184 nM respectively) but not SK-MEL-28 skin melanoma cells (IC50 10000 nM)
Catalog No. 50-294-8477