Learn More
Apexbio Technology LLC egg white lysozyme (19-36) [Gallus gallus] 5mg
SDP
Supplier:  Apexbio Technology LLC A10655
Encompass_Preferred
Egg white lysozyme (19-36) [Gallus gallus] is a peptide fragment composed of the amino acid sequence KVFGRCELAAAMKRHGLD derived from residues 19-36 of hen egg-white lysozyme Its molecular formula is C H N O S Lysozyme enzymatically hydrolyzes -(1-4) glycosidic linkages between N-acetylmuramic acid and N-acetylglucosamine residues within bacterial cell wall polysaccharides causing targeted cell lysis This fragment retains structural characteristics relevant to the parent enzyme s function It serves as a biochemical research tool for investigating bacterial cell wall integrity enzyme-substrate interaction mechanisms and the structural determinants underlying glycosidase activity in biomedical and microbiological research contexts
Catalog No. 50-000-63067