Learn More
Medchemexpress LLC EGGGG-PEG8-amide-bis(deoxyglucitol) | 2842855-38-1 | 98.90% | C49H91N9O28 | 5 MG

Supplier: Medchemexpress LLC HY494845MG
EGGGG-PEG8-amide-bis(deoxyglucitol) is characterized as a cleavable ADC (Antibody-Drug Conjugate) linker. It appears as a solid-liquid mixture, white to off-white in color, with a molecular weight of 1254.29 and the chemical formula C49H91N9O28. This compound is intended for research use only.
- Cleavable ADC linker: functions as a component in antibody-drug conjugates, facilitating the release of a therapeutic payload at the target site.
- Appearance: presented as a white to off-white solid-liquid mixture.
- Solubility: soluble in DMSO at a concentration of 75 mg/mL (59.79 mM) with the aid of ultrasonication and warming to 60°C.
- Storage conditions: solid form: store at -20°C, protected from light. In solvent (stock solution): stable for 6 months at -80°C and 1 month at -20°C, protected from light.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.