Learn More
Medchemexpress LLC Eglu | 170984-72-2 | 99.8% | 175.18 | C7H13NO4 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY1013321MG
Encompass_Preferred
EGLU ((2S)-α-ethylglutamic acid) is a selective, competitive antagonist of metabotropic glutamate receptor 2 (mGluR2) used as a small-molecule research reagent in neuropharmacology. Supplied as a purified solid for laboratory research applications.
  • Selective mGluR2 antagonist for receptor pharmacology studies.
  • High purity (99.8%) for reliable experimental results.
  • Molecular formula C7H13NO4 and molecular weight 175.18.
  • Available in milligram-scale quantities suitable for small-scale assays.
  • Characterized solid with supporting datasheet and safety information.
Catalog No. 50-002-12812