Learn More
Medchemexpress LLC Eglumegad hydrochloride | 176199-48-7 | 99.63% | 221.64 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY18941C5MG
Encompass_Preferred
Eglumegad hydrochloride is a highly potent and selective group II (mGlu2/3) receptor agonist. It protects neurons from NMDA toxicity and has anxiolytic- and antipsychotic-like effects.
  • Highly potent and selective group II (mGlu2/3) receptor agonist.
  • Protects neurons from NMDA toxicity.
  • Has anxiolytic-like effects.
  • Has antipsychotic-like effects.
Catalog No. 50-003-85085