Learn More
Selleck Chemical LLC EGNHS E7189-50MG
SDP
Supplier:  Selleck Chemical LLC E718950MG
Encompass_Preferred
EGNHS is a proteolytic targeting chimera (PROTAC) linker belonging to the alkyl/ether class used in the synthesis of PROTAC molecules It can form a cross-link between Lys232 and Lys241 of horseradish peroxidase C (HRP) and modify Lys174 without cross-linking
Catalog No. 50-000-43709