Learn More
Medchemexpress LLC Egnhs | 70539-42-3 | >97.5% | 250 MG

Supplier: Medchemexpress LLC 250MGHY130458
EGNHS is an alkyl/ether-based PROTAC linker utilized in the synthesis of PROTACs. PROTACs, or Proteolysis-Targeting Chimeras, consist of two different ligands connected by a linker; one ligand targets an E3 ubiquitin ligase, and the other targets the protein of interest. This mechanism exploits the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
- Alkyl/ether-based PROTAC linker
- Used in the synthesis of PROTACs
- Exploits intracellular ubiquitin-proteasome system to selectively degrade target proteins
- Storage as powder: -20°C for 3 years, 4°C for 2 years
- Storage in solvent: -80°C for 6 months, -20°C for 1 month
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.