Learn More
Enzo Life Sciences EGTA (2x) Buffer (1ml)
SDP
Supplier:  Enzo Life Sciences BMLKI1360001
Encompass_Preferred
Formulation: Liquid. 100mM TRIS, pH 7.5, containing 20mM EGTA, 200mM sodium chloride, 12mM magnesium chloride, 1mM DTT and 0.05% NP-40. Long Term Storage: -80°C.
Catalog No. 50-201-1093