Learn More
Medchemexpress LLC EGTA | 67-42-5 | 98.0% | 1 G
SDP
Supplier:  Medchemexpress LLC HYD08611G
Encompass_Preferred
EGTA is a cell-impermeant specific calcium ion chelator. It has an apparent calcium dissociation constant (Kd) of 60.5 nM at physiological pH (7.4) and exhibits very high specificity for Ca 2+ over Mg 2+ (Mg 2+ Kd 1-10 mM). EGTA significantly inhibits the substrate adherence capacity of inflammatory macrophages.
  • Cell-impermeant specific calcium ion chelator.
  • Apparent calcium dissociation constant (Kd) of 60.5 nM at physiological pH (7.4).
  • Very high specificity for Ca 2+ over Mg 2+ (Mg 2+ Kd 1-10 mM).
  • Significantly inhibits the substrate adherence capacity of inflammatory macrophages.
Catalog No. 50-004-09989