Learn More
Selleck Chemical LLC EGTA-AM E7574-1MG
SDP
Supplier:  Selleck Chemical LLC E75741MG
Encompass_Preferred
EGTA-AM is a membrane permeable form of EGTA can be passively loaded into cells to generate intracellular EGTA EGTA-AM is also a Ca2 chelator with slow chelating dynamics
Catalog No. 50-000-43455