Learn More
INTACT GENOMICS INC Thymidine Auxotrophic EHA105 Agrobacterium Chemically Competent Cells 5x50 µL
Supplier:  INTACT GENOMICS INC 130405
Encompass
Intact Genomics (ig) Thymidine Auxotrophic EHA105Thy Agrobacterium Chemically Competent Cells include modifications so that they will not grow unless 50 mg/L of thymidine is added to Minimal medium. This prevents the bacteria from overgrowing plant tissues when used for plant transformation. Thymidine-Auxotrophic EHA105Thy Agrobacterium Chemically Competent Cells are optimized for the highest transformation efficiencies. The EHA105Thy strain is useful for transgenic operations of rice, tobacco and other plants. EAH105Thy– thyA knockout mutant derived from EHA105, itself a derivative of A281 (A136/pTiBo542).Benefits: Thymidine Auxotrophic Reduced bacterial overgrowth during co-cultivation Decreased need for antibioticsProduct Components: Agrobacterium Chemically Competent Cells DNA (pCAMBIA1391z, 500 pg/µl) Recovery medium* * Client needs to add 50 mg/L of thymidine (not provided) to recovery media before use
Catalog No. 50-000-82952