Learn More
Creative Bioarray CSC-C3439, EHEB, Species: Human, Source: chronic B cell leukemia
Supplier:  Creative Bioarray CSCC3439
Encompass
Established from the peripheral blood of a 69-year-old woman with B-CLL (chronic lymphocytic leukemia) prior treatment by EBV-transformation in 1988; cell line may represent rather a B-lymphoblastoid cell line than a bona fide B-CLL cell line
Catalog No. NC2961553