Learn More
Sino Biological Recombinant Human DNMT3L Protein (Full Length, GST Tag), Active
Supplier:  Sino Biological D353380CG50
Encompass_Preferred
Recombinant full length human DNMT3L was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
Catalog No. 50-257-5319