Learn More
Medchemexpress LLC EHNA hydrochloride | 58337-38-5 | MFCD00151066 | 99.7% | 313.83 g/mol | C14H24ClN5O | 1 ML

Supplier: Medchemexpress LLC HY103160A10MM 1ML
EHNA hydrochloride is the hydrochloride salt of erythro-9-(2-hydroxy-3-nonyl)adenine, a selective phosphodiesterase (PDE) inhibitor and adenosine deaminase inhibitor used in biochemical research. It is provided as a 10 mM solution in DMSO for direct use in in vitro assays and signaling studies.
- High purity: 99.67%.
- Molecular weight 313.83 g/mol; formula C14H24ClN5O.
- Supplied as a 10 mM solution in DMSO (1 mL).
- Solubility: readily soluble in DMSO and water under recommended conditions.
- Storage: solid at -20°C; solution recommended at -80°C for long-term storage.
- For research use only; not for human or diagnostic use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.