Learn More
Medchemexpress LLC HY-12810 10mg Medchemexpress, EHop-016 CAS:1380432-32-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1281010MG
Encompass_Preferred
Medchemexpress, HY-12810 10mg EHop-016 CAS:1380432-32-5 EHop-016 is a potent and selective Rac GTPase Rac1 and Rac3 inhibitor. EHop-016 inhibits Rac1 activity with an IC50 of 1.1 μM in MDA-MB-435 cells. EHop-016 inhibits Vav2 interaction with Rac, Rac-activated PAK1, lamellipodia formation, and cell migration. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1150