Learn More
Selleck Chemical LLC EHT 1864 2HCl 10mg 754240-09-0
SDP
Supplier:  Selleck Chemical LLC S748210MG
Encompass_Preferred
EHT 1864 2HCl is a potent Rac family GTPase inhibitor with Kd of 40 nM, 50 nM, 60 nM and 250 nM for Rac1, Rac1b, Rac2 and Rac3, respectively. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-4831